Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004979.1 | NP_001004979.1 | 1 - 2 | 728 | 61 | |
XM_012964530.2 | XP_012819984.1 | 1 - 2 | 924 | 61 | ||
GenBank mRNA | BC075522.1 | - | 1 - 2 | 728 | 61 | |
CR760349.2 | - | 1 - 2 | 790 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |