Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005026.1 | NP_001005026.1 | 2 - 3 | 159 | 93 | |
XM_018091168.1 | XP_017946657.1 | 3 - 4 | 216 | 93 | ||
GenBank mRNA | BC076707.1 | - | 2 - 3 | 159 | 93 | |
BC157264.1 | - | 1395 | 93 | |||
CT030557.1 | - | 2 - 3 | 157 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |