Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203521.1 | NP_988852.1 | 5 - 6 | 899 | 66 | |
GenBank mRNA | AY079194.1 | - | 5 - 6 | 899 | 66 | |
CR762022.2 | - | 5 - 6 | 871 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |