Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203545.1 | NP_988876.1 | 5 - 6 | 588 | 88 | |
XM_012958022.2 | XP_012813476.1 | 5 - 6 | 639 | 88 | ||
XM_012958023.2 | XP_012813477.1 | 6 - 7 | 811 | 88 | ||
GenBank mRNA | BC059737.1 | - | 5 - 6 | 588 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |