Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203619.1 | NP_988950.1 | 2 - 3 | 934 | 95 | |
GenBank mRNA | BC061325.1 | - | 2 - 3 | 934 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |