Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203634.1 | NP_988965.1 | 7 - 8 | 999 | 95 | |
GenBank mRNA | BC061429.1 | - | 7 - 8 | 999 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |