Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203659.1 | NP_988990.1 | 1 - 2 | 229 | 97 | |
GenBank mRNA | BC061301.1 | - | 1 - 2 | 229 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |