Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203716.1 | NP_989047.1 | 5 - 6 | 622 | 91 | |
XM_012962836.2 | XP_012818290.1 | 5 - 6 | 757 | 91 | ||
GenBank mRNA | BC061354.1 | - | 5 - 6 | 622 | 91 | |
CR761467.2 | - | 5 - 6 | 489 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |