Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203730.2 | NP_989061.2 | 7 - 8 | 1200 | 67 | |
GenBank mRNA | BC061314.1 | - | 7 - 8 | 1192 | 67 | |
CX877195.2 | - | 1 - 2 | 669 | 67 | ||
DN018992.1 | - | 1 - 2 | 651 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |