Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203853.1 | NP_989184.1 | 2 - 3 | 315 | 95 | |
XM_012964435.2 | XP_012819889.1 | 2 - 3 | 381 | 95 | ||
GenBank mRNA | BC063209.1 | - | 2 - 3 | 315 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |