Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tktl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203896.1 | NP_989227.1 | 13 - 14 | 1752 | 93 | |
GenBank mRNA | BC063226.1 | - | 13 - 14 | 1752 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |