Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203933.1 | NP_989264.1 | 1 - 2 | 202 | 70 | |
GenBank mRNA | BC064247.1 | - | 1 - 2 | 202 | 70 | |
BC080335.1 | - | 1 - 2 | 215 | 70 | ||
CR760236.2 | - | 1 - 2 | 153 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |