Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203958.1 | NP_989289.1 | 7 - 8 | 981 | 78 | |
GenBank mRNA | BC064175.1 | - | 7 - 8 | 981 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |