Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204005.1 | NP_989336.1 | 3 - 4 | 417 | 78 | |
GenBank mRNA | AY490096.1 | - | 3 - 4 | 485 | 78 | |
BC064192.1 | - | 3 - 4 | 417 | 78 | ||
CR855790.2 | - | 3 - 4 | 407 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |