Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005131.1 | NP_001005131.1 | 1 - 2 | 234 | 84 | |
XM_012960204.2 | XP_012815658.1 | 1 - 2 | 353 | 84 | ||
GenBank mRNA | BC077669.1 | - | 1 - 2 | 234 | 84 | |
CR760027.2 | - | 1 - 2 | 97 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |