Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005131.1 | NP_001005131.1 | 3 - 4 | 439 | 83 | |
XM_012960204.2 | XP_012815658.1 | 3 - 4 | 558 | 83 | ||
GenBank mRNA | BC077669.1 | - | 3 - 4 | 439 | 83 | |
CR760027.2 | - | 3 - 4 | 302 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |