Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cops9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005143.1 | NP_001005143.1 | 1 - 2 | 155 | 66 | |
GenBank mRNA | BC077691.1 | - | 1 - 2 | 155 | 66 | |
CR761705.2 | - | 1 - 2 | 90 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |