Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005644.1 | NP_001005644.1 | 3 - 4 | 783 | 86 | |
XM_012958805.2 | XP_012814259.1 | 4 - 5 | 879 | 86 | ||
XM_012958806.1 | XP_012814260.1 | 4 - 5 | 778 | 86 | ||
GenBank mRNA | BC074644.1 | - | 3 - 4 | 783 | 86 | |
CR942435.3 | - | 3 - 4 | 528 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |