Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005817.1 | NP_001005817.1 | 5 - 6 | 608 | 76 | |
XM_012960840.2 | XP_012816294.1 | 5 - 6 | 650 | 76 | ||
XM_012960841.2 | XP_012816295.1 | 5 - 6 | 596 | 76 | ||
GenBank mRNA | BC075366.1 | - | 5 - 6 | 608 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |