Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006766.1 | NP_001006767.1 | 2 - 3 | 212 | 83 | |
XM_012958845.2 | XP_012814299.1 | 2 - 3 | 686 | 83 | ||
GenBank mRNA | BC075560.1 | - | 2 - 3 | 212 | 83 | |
CR760740.2 | - | 1 - 2 | 102 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |