Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006768.1 | NP_001006769.1 | 7 - 8 | 646 | 85 | |
GenBank mRNA | BC075563.1 | - | 7 - 8 | 646 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |