Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006880.2 | NP_001006881.1 | 2 - 3 | 217 | 91 | |
XM_012960188.2 | XP_012815642.1 | 3 - 4 | 302 | 91 | ||
XM_018091175.1 | XP_017946664.1 | 3 - 4 | 304 | 91 | ||
GenBank mRNA | BC077045.1 | - | 2 - 3 | 218 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |