Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006889.2 | NP_001006890.1 | 5 - 6 | 383 | 85 | |
XM_012960882.2 | XP_012816336.1 | 5 - 6 | 85 | |||
GenBank mRNA | BC077689.1 | - | 5 - 6 | 385 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |