Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006917.1 | NP_001006918.1 | 1 - 2 | 212 | 89 | |
GenBank mRNA | BC075503.1 | - | 1 - 2 | 212 | 89 | |
CR761169.2 | - | 1 - 2 | 146 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |