Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: naa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007496.1 | NP_001007497.1 | 6 - 7 | 421 | 81 | |
XM_012968260.2 | XP_012823714.1 | 6 - 7 | 490 | 81 | ||
GenBank mRNA | BC079933.1 | - | 6 - 7 | 421 | 81 | |
CR760841.2 | - | 6 - 7 | 423 | 81 | ||
CR848272.2 | - | 6 - 7 | 390 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |