Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007513.1 | NP_001007514.1 | 11 - 12 | 1672 | 91 | |
XM_012958864.1 | XP_012814318.1 | 11 - 12 | 1734 | 91 | ||
XM_012958865.1 | XP_012814319.1 | 11 - 12 | 1731 | 91 | ||
XM_012958867.2 | XP_012814321.1 | 11 - 12 | 1714 | 91 | ||
XM_012958868.2 | XP_012814322.1 | 11 - 12 | 1704 | 91 | ||
XM_012958869.1 | XP_012814323.1 | 11 - 12 | 1531 | 91 | ||
XM_012958870.2 | XP_012814324.1 | 11 - 12 | 1548 | 91 | ||
XM_012958871.2 | XP_012814325.1 | 11 - 12 | 1533 | 91 | ||
GenBank mRNA | BC079954.1 | - | 11 - 12 | 1672 | 91 | |
CT030587.1 | - | 11 - 12 | 1513 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |