Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mturn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007922.2 | NP_001007923.2 | 2 - 3 | 551 | 106 | |
GenBank mRNA | BC080370.1 | - | 2 - 3 | 450 | 106 | |
BX731725.1 | - | 2 - 3 | 551 | 106 | ||
CR761610.2 | - | 2 - 3 | 466 | 106 | ||
CR926360.1 | - | 2 - 3 | 532 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |