Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007945.2 | NP_001007946.1 | 4 - 5 | 560 | 86 | |
GenBank mRNA | BC080452.1 | - | 4 - 5 | 561 | 86 | |
CR760810.2 | - | 4 - 5 | 449 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |