Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007957.1 | NP_001007958.1 | 4 - 5 | 577 | 67 | |
GenBank mRNA | BC080471.1 | - | 4 - 5 | 577 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |