Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007990.1 | NP_001007991.1 | 4 - 5 | 412 | 100 | |
XM_012953584.2 | XP_012809038.1 | 4 - 5 | 500 | 100 | ||
XM_012953585.2 | XP_012809039.1 | 4 - 5 | 511 | 100 | ||
GenBank mRNA | BC080503.1 | - | 4 - 5 | 412 | 100 | |
CR926422.2 | - | 3 - 4 | 352 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |