Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007999.1 | NP_001008000.1 | 411 | 65 | ||
GenBank mRNA | BC080878.1 | - | 411 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |