Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008039.1 | NP_001008040.1 | 6 - 7 | 1060 | 67 | |
GenBank mRNA | BC080923.1 | - | 6 - 7 | 1060 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |