Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008112.1 | NP_001008113.1 | 3 - 4 | 1774 | 82 | |
GenBank mRNA | BC081317.1 | - | 3 - 4 | 1774 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |