Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008119.1 | NP_001008120.1 | 1388 | 127 | ||
GenBank mRNA | BC081324.1 | - | 1388 | 127 | ||
CR760728.2 | - | 1309 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis |