Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008123.1 | NP_001008124.1 | 7 - 8 | 696 | 98 | |
XM_012970344.2 | XP_012825798.1 | 7 - 8 | 792 | 98 | ||
XM_012970345.2 | XP_012825799.1 | 6 - 7 | 746 | 98 | ||
XM_012970346.1 | XP_012825800.1 | 6 - 7 | 691 | 98 | ||
GenBank mRNA | BC081329.1 | - | 7 - 8 | 696 | 98 | |
CR761020.2 | - | 6 - 7 | 593 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |