Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008154.2 | NP_001008155.1 | 6 - 7 | 729 | 83 | |
XM_012966478.2 | XP_012821932.1 | 5 - 6 | 791 | 83 | ||
XM_012966479.2 | XP_012821933.1 | 6 - 7 | 822 | 83 | ||
GenBank mRNA | BC081368.1 | - | 6 - 7 | 731 | 83 | |
CR762026.2 | - | 5 - 6 | 702 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |