Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008169.1 | NP_001008170.1 | 7 - 8 | 1070 | 63 | |
XM_018093863.1 | XP_017949352.1 | 7 - 8 | 1007 | 63 | ||
XM_018093864.1 | XP_017949353.1 | 7 - 8 | 1121 | 63 | ||
GenBank mRNA | BC082488.1 | - | 7 - 8 | 1070 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |