Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008187.1 | NP_001008188.1 | 5 - 6 | 789 | 93 | |
GenBank mRNA | BC082513.1 | - | 5 - 6 | 789 | 93 | |
CU075423.1 | - | 5 - 6 | 790 | 93 |
Gene Symbol: LOC100498595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002940779.4 | XP_002940825.2 | 4 - 5 | 1065 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |