Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011071.1 | NP_001011071.1 | 3 - 4 | 374 | 55 | |
XM_012953486.2 | XP_012808940.1 | 3 - 4 | 355 | 55 | ||
GenBank mRNA | BC084454.1 | - | 3 - 4 | 374 | 55 | |
CR761781.2 | - | 2 - 3 | 282 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |