Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011091.1 | NP_001011091.1 | 1 - 2 | 617 | 90 | |
GenBank mRNA | BC084483.1 | - | 1 - 2 | 617 | 90 | |
CT030281.1 | - | 1 - 2 | 612 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |