Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011169.1 | NP_001011169.1 | 561 | 66 | ||
GenBank mRNA | BC084995.1 | - | 561 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |