Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dctn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011198.2 | NP_001011198.1 | 4 - 5 | 369 | 106 | |
GenBank mRNA | BC087561.2 | - | 4 - 5 | 369 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |