Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011201.2 | NP_001011201.1 | 5 - 6 | 656 | 100 | |
GenBank mRNA | BC087571.2 | - | 5 - 6 | 656 | 100 | |
CT030432.1 | - | 5 - 6 | 719 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |