Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011363.1 | NP_001011363.1 | 4 - 5 | 668 | 86 | |
XM_012956865.2 | XP_012812319.1 | 3 - 4 | 577 | 86 | ||
XM_012956867.2 | XP_012812321.1 | 3 - 4 | 535 | 86 | ||
GenBank mRNA | BC088529.1 | - | 4 - 5 | 668 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |