Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015899.1 | NP_001015899.1 | 2 - 3 | 231 | 76 | |
XM_012961035.2 | XP_012816489.1 | 1 - 2 | 1484 | 76 | ||
GenBank mRNA | BC118739.1 | - | 2 - 3 | 213 | 76 | |
CR926447.1 | - | 2 - 3 | 231 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |