Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015912.1 | NP_001015912.1 | 3 - 4 | 431 | 101 | |
GenBank mRNA | BC135957.1 | - | 3 - 4 | 453 | 101 | |
CR926424.1 | - | 3 - 4 | 431 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |