Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016133.2 | NP_001016133.1 | 8 - 9 | 1281 | 92 | |
XM_018093063.1 | XP_017948552.1 | 7 - 8 | 2286 | 92 | ||
GenBank mRNA | BC090614.1 | - | 8 - 9 | 1281 | 92 | |
CR762041.2 | - | 9 - 10 | 1171 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |