Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gstp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016641.2 | NP_001016641.1 | 5 - 6 | 383 | 73 | |
GenBank mRNA | BC135948.1 | - | 5 - 6 | 387 | 73 | |
CR760401.2 | - | 5 - 6 | 383 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |