Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016676.2 | NP_001016676.1 | 5 - 6 | 465 | 85 | |
XM_018093890.1 | XP_017949379.1 | 6 - 7 | 626 | 85 | ||
GenBank mRNA | BC170614.1 | - | 5 - 6 | 463 | 85 | |
BC170620.1 | - | 5 - 6 | 463 | 85 | ||
CR760229.2 | - | 5 - 6 | 465 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |