Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016743.1 | NP_001016743.1 | 1 - 2 | 699 | 87 | |
GenBank mRNA | CR759991.1 | - | 1 - 2 | 699 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |